Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281295.1 | NP_001268224.1 | 4 | 642 | ||
NM_001281296.1 | NP_001268225.1 | 4 | 642 | |||
NM_001321739.1 | NP_001308668.1 | 4 | 642 | |||
NM_138804.4 | NP_620159.2 | 4 | 633 | |||
XM_006711946.3 | XP_006712009.1 | 4 | 978 | |||
XM_011532548.2 | XP_011530850.1 | 4 | 979 | |||
XM_011532549.2 | XP_011530851.1 | 4 | 980 | |||
XM_011532550.2 | XP_011530852.1 | 4 | 589 | |||
XM_011532551.2 | XP_011530853.1 | 4 | 588 | |||
XM_011532552.2 | XP_011530854.1 | 4 | 979 | |||
XM_011532553.2 | XP_011530855.1 | 4 | 980 | |||
GenBank mRNA | AK300188.1 | - | 4 | 634 | ||
BC014602.2 | - | 4 | 611 | 4821635 | ||
BC025997.1 | - | 4 | 636 | 4823281 |