Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122870.2 | NP_001116342.1 | 4, 5 | 459 | ||
NM_144641.3 | NP_653242.3 | 5, 6 | 825 | |||
XM_005264879.2 | XP_005264936.1 | 5, 6 | 893 | |||
XM_005264880.3 | XP_005264937.1 | 4, 5 | 638 | |||
XM_005264881.1 | XP_005264938.1 | 5, 6 | 714 | |||
GenBank mRNA | AB474372.1 | - | 5, 6 | 793 | ||
AK056894.1 | - | 5, 6 | 714 | |||
BC009644.1 | - | 4, 5 | 420 | 3905720 |