Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115007.2 | NP_001108479.1 | 14 | 1602 | ||
NM_001115008.2 | NP_001108480.1 | 14 | 1803 | |||
NM_001288996.1 | NP_001275925.1 | 11 | 2199 | |||
NM_001288997.1 | NP_001275926.1 | 14 | 1540 | |||
NM_194282.3 | NP_919258.2 | 14 | 2466 | |||
NR_110253.1 | 11 | 2125 | ||||
NR_110254.1 | 13 | 1799 | ||||
XM_005262750.4 | XP_005262807.1 | 13 | 2275 | |||
XM_006714081.3 | XP_006714144.1 | 13 | 2201 | |||
XM_017007728.1 | XP_016863217.1 | 11 | 2008 | |||
XM_017007729.1 | XP_016863218.1 | 12 | 2191 | |||
XM_017007730.1 | XP_016863219.1 | 12 | 1580 | |||
GenBank mRNA | AB111889.1 | - | 13 | 1786 | ||
AK292769.1 | - | 14 | 1550 | |||
AK294256.1 | - | 14 | 2395 | |||
BC109277.2 | - | 11 | 2001 | |||
BC109278.2 | - | 11 | 2074 | 40028371 | ||
BX537919.1 | - | 13 | 1700 | |||
BX640657.1 | - | 14 | 1529 | |||
BX640966.1 | - | 14 | 2461 |