Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDUFAF6 SIRNA Assays ›
          Gene Symbol:
          NDUFAF6
          Gene Name:
          NADH:ubiquinone oxidoreductase complex assembly factor 6
          Gene Aliases:
          C8orf38
          Chromosome Location:
          Chr.8: 94895271 - 95118496 on Build GRCh38
          UniGene ID:
          Hs.729144
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS134674
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NDUFAF6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_152416.3 NP_689629.2 1, 2 230
          XM_005250789.2 XP_005250846.1 1, 2 237
          XM_005250790.2 XP_005250847.1 1, 2 222
          XM_005250791.2 XP_005250848.1 1, 2 183
          XM_005250792.2 XP_005250849.1 1, 2 183
          XM_005250793.2 XP_005250850.1 1, 2 27
          XM_011516833.2 XP_011515135.1 6, 7 1450
          XM_011516834.2 XP_011515136.1 5, 6 1411
          XM_011516835.2 XP_011515137.1 5, 6 669
          XM_011516836.2 XP_011515138.1 5, 6 1352
          XM_011516837.2 XP_011515139.1 6, 7 1410
          XM_011516838.2 XP_011515140.1 5, 6 1287
          XM_011516839.2 XP_011515141.1 4, 5 1189
          XM_011516840.2 XP_011515142.1 5, 6 1283
          XM_011516841.2 XP_011515143.1 5, 6 1312
          XM_011516842.2 XP_011515144.1 4, 5 1185
          XM_017013027.1 XP_016868516.1 5, 6 661
          XM_017013028.1 XP_016868517.1 4, 5 561
          XM_017013029.1 XP_016868518.1 3, 4 1150
          XM_017013030.1 XP_016868519.1 3, 4 895
          XM_017013031.1 XP_016868520.1 4, 5 715
          XM_017013032.1 XP_016868521.1 2, 3 337
          XM_017013033.1 XP_016868522.1 1, 2 34
          XM_017013035.1 XP_016868524.1 3, 4 566
          XM_017013036.1 XP_016868525.1 3, 4 971
          XM_017013037.1 XP_016868526.1 1, 2 28
          XM_017013038.1 XP_016868527.1 1, 2 60
          XM_017013039.1 XP_016868528.1 2, 3 447
          GenBank mRNA AK298631.1 - 1, 2 150
          AK301414.1 - 1, 2 148
          AY444560.1 - 2, 3 301
          BC028166.1 - 1, 2 222
          BM755491.1 - 1, 2 230
          BP275471.1 - 1, 2 223

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biosynthetic process
          mitochondrial respiratory chain complex I assembly
          transmembrane transport
          transferase activity
          transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0