Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALYL SIRNA Assays ›
Gene Symbol:
RALYL
Gene Name:
RALY RNA binding protein-like
Gene Aliases:
HNRPCL3
Chromosome Location:
Chr.8: 84182629 - 84921844 on Build GRCh38
UniGene ID:
Hs.121663
Species:
Human
siRNA ID
HSS134708
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALYL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100391.1
NP_001093861.1
7
830
NM_001100392.1
NP_001093862.1
8
1073
NM_001100393.1
NP_001093863.1
7
1041
NM_001287243.1
NP_001274172.1
6
1721
NM_001287244.1
NP_001274173.1
6
535
NM_173848.5
NP_776247.3
7
1754
XM_011517462.2
XP_011515764.1
7
1825
XM_017013058.1
XP_016868547.1
6
996
XM_017013059.1
XP_016868548.1
9
1124
XM_017013060.1
XP_016868549.1
7
3274
XM_017013061.1
XP_016868550.1
7
2426
XM_017013062.1
XP_016868551.1
9
1233
XM_017013063.1
XP_016868552.1
7
996
XM_017013064.1
XP_016868553.1
8
1347
XM_017013065.1
XP_016868554.1
7
895
XM_017013066.1
XP_016868555.1
6
1008
XM_017013067.1
XP_016868556.1
7
1899
XM_017013068.1
XP_016868557.1
8
1091
XM_017013069.1
XP_016868558.1
8
1147
XM_017013070.1
XP_016868559.1
7
991
XM_017013071.1
XP_016868560.1
6
862
XM_017013072.1
XP_016868561.1
7
982
XM_017013073.1
XP_016868562.1
7
982
XM_017013074.1
XP_016868563.1
7
895
XM_017013075.1
XP_016868564.1
6
947
XM_017013076.1
XP_016868565.1
7
775
XM_017013077.1
XP_016868566.1
8
2025
XM_017013078.1
XP_016868567.1
7
1041
XM_017013079.1
XP_016868568.1
8
1024
XM_017013080.1
XP_016868569.1
6
742
XM_017013081.1
XP_016868570.1
6
742
XM_017013082.1
XP_016868571.1
6
1008
XM_017013083.1
XP_016868572.1
6
696
XM_017013084.1
XP_016868573.1
5
1531
XM_017013085.1
XP_016868574.1
7
578
XM_017013086.1
XP_016868575.1
6
535
XM_017013087.1
XP_016868576.1
6
535
GenBank mRNA
AF070623.1
-
1
77
AF495529.1
-
7
1754
AF495530.1
-
7
971
AK094543.1
-
7
985
AK095026.1
-
6
1537
AK095560.1
-
8
1024
AK123437.1
-
6
535
BC031090.1
-
7
819
5311935
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0