Search Thermo Fisher Scientific
Gene Symbol: MUC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040105.1 | NP_001035194.1 | 3 | 10774 | ||
NR_133665.1 | 3 | 10774 | ||||
GenBank mRNA | AF016692.1 | - | 1 | 331 | ||
AF430017.1 | - | 1 | 331 | |||
AJ606307.1 | - | 3 | 10774 | |||
AJ606308.1 | - | 3 | 10774 | |||
BC126315.1 | - | 1 | 916 |