Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040105.1 | NP_001035194.1 | 3 | 501 | ||
NR_133665.1 | 3 | 501 | ||||
GenBank mRNA | AJ606307.1 | - | 3 | 501 | ||
AJ606308.1 | - | 3 | 501 |