Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138477.2 | NP_612486.2 | 22 | 3028 | ||
XM_005254176.4 | XP_005254233.1 | 22 | 2943 | |||
XM_011521270.2 | XP_011519572.1 | 22 | 2968 | |||
XM_011521271.2 | XP_011519573.1 | 22 | 2965 | |||
XM_011521274.2 | XP_011519576.1 | 19 | 2014 | |||
GenBank mRNA | AF525398.1 | - | 22 | 3025 | ||
BC001092.2 | - | 14 | 1483 | |||
BC008333.1 | - | 14 | 1483 | |||
BC008334.1 | - | 14 | 1483 | |||
BC052568.1 | - | 22 | 2916 | |||
BC066640.1 | - | 22 | 2916 | 6577335 |