Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145303.2 | NP_001138775.2 | 13 | 2031 | ||
NM_144686.3 | NP_653287.2 | 13 | 2013 | |||
XM_011526486.2 | XP_011524788.1 | 10 | 1516 | |||
GenBank mRNA | AK314973.1 | - | 13 | 1934 | ||
AY236492.1 | - | 13 | 2030 | |||
AY263166.1 | - | 13 | 1910 | |||
BC025323.1 | - | 13 | 1920 | 2959382 | ||
BC032477.1 | - | 7 | 1121 | |||
BU680152.1 | - | 2 | 142 |