Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 3, 4 | 446 | ||
NM_207325.2 | NP_997208.2 | 3, 4 | 472 | |||
XM_011526526.2 | XP_011524828.1 | 3, 4 | 427 | |||
XM_017026366.1 | XP_016881855.1 | 3, 4 | 463 | |||
XM_017026367.1 | XP_016881856.1 | 3, 4 | 430 | |||
GenBank mRNA | AF533710.1 | - | 3, 4 | 398 | ||
AK126757.1 | - | 3, 4 | 463 | |||
AK129497.1 | - | 3, 4 | 427 | |||
BC131725.1 | - | 2, 3 | 294 | 40123868 |