Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152494.3 | NP_689707.2 | 3, 4 | 300 | ||
GenBank mRNA | AK057347.1 | - | 3, 4 | 282 | ||
AK301902.1 | - | 3, 4 | 379 | |||
BC064844.1 | - | 2, 3 | 255 | 30378702 |