Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABCA13 SIRNA Assays ›
Gene Symbol:
ABCA13
Gene Name:
ATP binding cassette subfamily A member 13
Gene Aliases:
-
Chromosome Location:
Chr.7: 48171424 - 48647497 on Build GRCh38
UniGene ID:
Hs.226568
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS135954
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABCA13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152701.4
NP_689914.3
18
8692
XM_011515130.2
XP_011513432.1
18
8724
XM_011515131.2
XP_011513433.1
18
8724
XM_011515132.2
XP_011513434.1
18
8724
XM_011515133.2
XP_011513435.1
18
8724
XM_011515134.2
XP_011513436.1
18
8724
XM_011515136.2
XP_011513438.2
13
8089
XM_011515137.2
XP_011513439.1
18
8724
XM_011515138.2
XP_011513440.1
18
8724
XM_011515139.2
XP_011513441.1
18
8724
XM_011515141.2
XP_011513443.1
18
8725
XM_011515142.2
XP_011513444.1
18
8725
XM_011515143.2
XP_011513445.1
18
8724
XM_011515144.2
XP_011513446.1
18
8724
XM_011515145.2
XP_011513447.1
18
8725
XM_011515146.2
XP_011513448.1
18
8725
XM_011515147.2
XP_011513449.1
18
8726
XM_011515148.2
XP_011513450.1
18
8727
XM_011515149.2
XP_011513451.1
18
8727
XM_017011767.1
XP_016867256.1
18
8724
XM_017011768.1
XP_016867257.1
18
8724
XM_017011769.1
XP_016867258.1
18
8728
GenBank mRNA
AY204751.1
-
18
8693
JF913488.1
-
1
587
JF913491.1
-
1
587
JF913492.1
-
1
587
Back To Top
More Information
Panther Classification:
Molecular Function -
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cholesterol efflux
phospholipid efflux
transmembrane transport
ATP binding
ATPase activity, coupled to transmembrane movement of substances
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0