Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168339.1 | NP_001161811.1 | 19 | 1968 | ||
NM_001168340.1 | NP_001161812.1 | 18 | 1935 | |||
NM_001168341.1 | NP_001161813.1 | 17 | 1767 | |||
NM_001168342.1 | NP_001161814.1 | 17 | 1849 | |||
NM_152574.2 | NP_689787.2 | 19 | 1974 | |||
XM_011517732.2 | XP_011516034.1 | 18 | 1771 | |||
XM_017014310.1 | XP_016869799.1 | 21 | 2337 | |||
XM_017014311.1 | XP_016869800.1 | 20 | 2269 | |||
XM_017014312.1 | XP_016869801.1 | 18 | 1839 | |||
GenBank mRNA | AK091187.1 | - | 19 | 1860 | ||
AK093459.1 | - | 19 | 2036 | |||
AK298581.1 | - | 17 | 1622 | |||
AK298998.1 | - | 18 | 1907 | |||
AK301427.1 | - | 17 | 1765 | |||
BC038592.1 | - | 6 | 806 | |||
BC042417.1 | - | 18 | 1841 | |||
BC142985.1 | - | 19 | 1947 | 8860428 |