Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145913.3 | NP_666018.3 | 11 | 1665 | ||
XM_017018910.1 | XP_016874399.1 | 11 | 1701 | |||
GenBank mRNA | AF536216.1 | - | 11 | 1638 | ||
AK075263.1 | - | 11 | 1323 | |||
AK313788.1 | - | 11 | 1634 | |||
AY081220.1 | - | 11 | 1639 | |||
BC110492.2 | - | 11 | 1300 | 40007567 |