Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198461.3 | NP_940863.3 | 3 | 1522 | ||
XM_017003491.1 | XP_016858980.1 | 3 | 1005 | |||
GenBank mRNA | AK127206.1 | - | 3 | 224 | ||
BC101662.1 | - | 2 | 181 | 8069168 | ||
BC101664.1 | - | 2 | 181 | 8069170 | ||
BC143468.1 | - | 2 | 181 | |||
DQ020495.1 | - | 3 | 1522 |