Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALMS1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003683.2 | 4 | 594 | |||
GenBank mRNA | AK307160.1 | - | 4 | 756 | ||
BC014492.1 | - | 4 | 594 |