Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC39A11 SIRNA Assays ›
Gene Symbol:
SLC39A11
Gene Name:
solute carrier family 39 member 11
Gene Aliases:
C17orf26, ZIP11
Chromosome Location:
Chr.17: 72645946 - 73092714 on Build GRCh38
UniGene ID:
Hs.221127
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS136966
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC39A11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159770.1
NP_001153242.1
5
462
NM_139177.3
NP_631916.2
5
462
XM_005257134.3
XP_005257191.1
5
549
XM_006721754.3
XP_006721817.1
5
549
XM_006721755.2
XP_006721818.1
6
753
XM_006721756.2
XP_006721819.1
5
462
XM_006721757.2
XP_006721820.1
5
462
XM_006721758.3
XP_006721821.1
5
462
XM_006721759.1
XP_006721822.1
5
462
XM_011524493.1
XP_011522795.1
6
844
XM_011524494.2
XP_011522796.1
5
495
XM_011524495.2
XP_011522797.1
5
462
XM_011524496.2
XP_011522798.1
5
462
XM_011524498.2
XP_011522800.1
5
462
XM_017024330.1
XP_016879819.1
6
753
XM_017024331.1
XP_016879820.1
6
844
XM_017024332.1
XP_016879821.1
5
459
XM_017024333.1
XP_016879822.1
4
402
XM_017024334.1
XP_016879823.1
5
549
XM_017024335.1
XP_016879824.1
5
462
XM_017024336.1
XP_016879825.1
6
642
XM_017024337.1
XP_016879826.1
6
555
XM_017024338.1
XP_016879827.1
5
462
XM_017024339.1
XP_016879828.1
5
462
XM_017024340.1
XP_016879829.1
5
462
XM_017024341.1
XP_016879830.1
5
549
XM_017024342.1
XP_016879831.1
5
462
GenBank mRNA
AF331643.1
-
5
424
AK313376.1
-
5
435
BC035631.1
-
5
441
5219031
DA380043.1
-
5
462
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
zinc II ion transmembrane transport
zinc ion transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0