Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STT3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178862.2 | NP_849193.1 | 6 | 1392 | ||
XM_011533465.1 | XP_011531767.1 | 6 | 1255 | |||
XM_017005857.1 | XP_016861346.1 | 6 | 1255 | |||
XM_017005858.1 | XP_016861347.1 | 6 | 713 | |||
XM_017005859.1 | XP_016861348.1 | 6 | 786 | |||
GenBank mRNA | AK074587.1 | - | 6 | 921 | ||
AK311488.1 | - | 6 | 1255 | |||
AX799082.1 | - | 6 | 894 | |||
AY074880.1 | - | 6 | 894 |