Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317078.1 | NP_001304007.1 | 2 | 557 | ||
NM_153450.2 | NP_703151.2 | 2 | 557 | |||
GenBank mRNA | AY148462.1 | - | 2 | 286 | ||
BC009723.1 | - | 2 | 433 | |||
BC037223.1 | - | 2 | 435 | 5197938 | ||
EB386171.1 | - | 2 | 443 | |||
HE646678.2 | - | 2 | 427 |