Search
Search

IL27| Gene Symbol: IL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145659.3 | NP_663634.2 | 3, 4 | 332 | ||
| XM_011545780.2 | XP_011544082.1 | 3, 4 | 686 | |||
| GenBank mRNA | AY099296.1 | - | 3, 4 | 309 | ||
| BC062422.1 | - | 3, 4 | 335 | 4703457 | ||