Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178863.4 | NP_849194.1 | 5 | 835 | ||
NR_110933.1 | 5 | 1373 | ||||
XM_011545783.2 | XP_011544085.1 | 5 | 904 | |||
XM_011545784.2 | XP_011544086.1 | 6 | 923 | |||
XM_017023104.1 | XP_016878593.1 | 6 | 869 | |||
GenBank mRNA | AF289573.1 | - | 5 | 782 | ||
AF401315.2 | - | 5 | 747 | |||
AK097407.1 | - | 5 | 978 | |||
AK289630.1 | - | 5 | 812 | |||
AY027918.1 | - | 5 | 759 | |||
BC036228.1 | - | 5 | 827 | 5276078 |