Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178510.1 | NP_848605.1 | 8 | 1825 | ||
XM_011542736.2 | XP_011541038.1 | 9 | 3635 | |||
XM_011542737.2 | XP_011541039.1 | 8 | 3605 | |||
XM_011542738.2 | XP_011541040.1 | 9 | 3893 | |||
XM_017017475.1 | XP_016872964.1 | 9 | 3632 | |||
GenBank mRNA | AF487542.1 | - | 8 | 1731 | ||
AJ541797.2 | - | 8 | 1825 |