Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101421.3 | NP_001094891.3 | 24 | 2475 | ||
XM_011538223.2 | XP_011536525.1 | 25 | 2983 | |||
XM_017019207.1 | XP_016874696.1 | 24 | 2917 | |||
GenBank mRNA | AK094906.1 | - | 1 | 246 | ||
AK298093.1 | - | 2 | 323 | |||
DC359165.1 | - | 2 | 324 |