Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203373.2 | NP_976307.2 | 1 | 359 | ||
XM_005254320.3 | XP_005254377.1 | 1 | 1487 | |||
XM_006720478.1 | XP_006720541.1 | 1 | 195 | |||
XM_011521468.2 | XP_011519770.1 | 1 | 325 | |||
XM_011521469.1 | XP_011519771.1 | 1 | 321 | |||
XM_011521472.2 | XP_011519774.1 | 1 | 325 | |||
XM_017022086.1 | XP_016877575.1 | 1 | 1487 | |||
GenBank mRNA | AA495872.1 | - | 1 | 360 | ||
AK096945.1 | - | 1 | 321 | |||
BC065833.1 | - | 1 | 315 | 30390268 |