Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENDOV SIRNA Assays ›
          Gene Symbol:
          ENDOV
          Gene Name:
          endonuclease V
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 80415117 - 80438086 on Build GRCh38
          UniGene ID:
          Hs.389678
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS138494
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ENDOV
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164637.2 NP_001158109.1 5 427
          NM_001164638.1 NP_001158110.1 5 427
          NM_173627.4 NP_775898.2 6 562
          XM_005257244.3 XP_005257301.1 6 612
          XM_005257246.3 XP_005257303.1 5 429
          XM_006721837.3 XP_006721900.1 6 612
          XM_011524655.1 XP_011522957.1 6 612
          XM_011524656.1 XP_011522958.1 6 612
          XM_011524657.1 XP_011522959.1 6 612
          XM_011524658.2 XP_011522960.1 6 612
          XM_011524660.1 XP_011522962.1 5 427
          XM_011524661.2 XP_011522963.1 5 429
          XM_011524662.1 XP_011522964.1 6 612
          XM_011524663.1 XP_011522965.1 6 612
          XM_011524664.2 XP_011522966.1 7 1230
          XM_011524666.1 XP_011522968.1 6 794
          XM_011524667.2 XP_011522969.1 6 740
          XM_011524669.2 XP_011522971.1 6 612
          XM_011524670.1 XP_011522972.1 6 612
          XM_011524671.2 XP_011522973.1 6 612
          XM_011524672.1 XP_011522974.1 5 410
          XM_011524673.1 XP_011522975.1 4 367
          XM_011524674.1 XP_011522976.1 5 410
          XM_011524675.1 XP_011522977.1 4 387
          XM_011524676.1 XP_011522978.1 5 659
          XM_011524677.1 XP_011522979.1 4 370
          XM_011524678.2 XP_011522980.1 4 525
          XM_017024508.1 XP_016879997.1 6 1559
          XM_017024509.1 XP_016879998.1 6 1505
          XM_017024510.1 XP_016879999.1 6 612
          XM_017024511.1 XP_016880000.1 5 429
          XM_017024512.1 XP_016880001.1 6 740
          XM_017024513.1 XP_016880002.1 3 485
          XM_017024514.1 XP_016880003.1 5 410
          XM_017024515.1 XP_016880004.1 4 370
          XM_017024516.1 XP_016880005.1 3 273
          XM_017024517.1 XP_016880006.1 4 387
          XM_017024518.1 XP_016880007.1 5 410
          XM_017024519.1 XP_016880008.1 4 370
          XM_017024520.1 XP_016880009.1 3 696
          GenBank mRNA AK092539.1 - 5 427
          AK096344.1 - 6 562
          AK096802.1 - 4 295
          AK123689.1 - 4 370
          BC037889.2 - 3 284
          BC045824.1 - 3 284
          BC064545.1 - 5 410 5736957
          BX647411.1 - 3 290

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          RNA phosphodiester bond hydrolysis, endonucleolytic
          magnesium ion binding
          single-stranded RNA binding
          endodeoxyribonuclease activity, producing 5'-phosphomonoesters
          endoribonuclease activity, producing 5'-phosphomonoesters
          structure-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0