Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173629.2 | NP_775900.1 | 2 | 229 | ||
XM_011525923.2 | XP_011524225.1 | 4 | 319 | |||
XM_011525924.2 | XP_011524226.1 | 4 | 347 | |||
XM_011525925.2 | XP_011524227.1 | 2 | 324 | |||
GenBank mRNA | AK096425.1 | - | 2 | 229 |