Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 20, 21 | 2562 | ||
NM_173636.4 | NP_775907.4 | 20, 21 | 2562 | |||
XM_005258809.2 | XP_005258866.1 | 20, 21 | 2562 | |||
XM_011526837.1 | XP_011525139.1 | 20, 21 | 2547 | |||
XM_011526838.1 | XP_011525140.1 | 20, 21 | 2562 | |||
XM_011526839.1 | XP_011525141.1 | 18, 19 | 2211 | |||
XM_011526840.2 | XP_011525142.1 | 13, 14 | 1557 | |||
XM_011526841.2 | XP_011525143.1 | 11, 12 | 1178 | |||
XM_011526842.1 | XP_011525144.1 | 10, 11 | 1010 | |||
XM_011526843.1 | XP_011525145.1 | 3, 4 | 337 | |||
XM_011526844.2 | XP_011525146.1 | 3, 4 | 334 | |||
XM_017026665.1 | XP_016882154.1 | 20, 21 | 2562 | |||
GenBank mRNA | BC017261.2 | - | 2, 3 | 271 | ||
BC058939.1 | - | 20, 21 | 2506 | |||
BQ880028.1 | - | 7, 8 | 731 | |||
BX647726.1 | - | 21, 22 | 2623 |