Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HKR1 SIRNA Assays ›
Gene Symbol:
HKR1
Gene Name:
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases:
ZNF875
Chromosome Location:
Chr.19: 37317900 - 37364455 on Build GRCh38
UniGene ID:
Hs.657027
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS138580
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HKR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181786.2
NP_861451.1
6
1955
XM_017026672.1
XP_016882161.1
4
2143
XM_017026673.1
XP_016882162.1
4
2209
XM_017026674.1
XP_016882163.1
8
2509
XM_017026675.1
XP_016882164.1
8
2506
XM_017026676.1
XP_016882165.1
5
1804
XM_017026677.1
XP_016882166.1
5
1791
XM_017026678.1
XP_016882167.1
5
1801
XM_017026679.1
XP_016882168.1
5
1788
XM_017026680.1
XP_016882169.1
8
2171
XM_017026681.1
XP_016882170.1
7
2119
XM_017026682.1
XP_016882171.1
8
2168
XM_017026683.1
XP_016882172.1
7
2030
XM_017026684.1
XP_016882173.1
6
1892
XM_017026685.1
XP_016882174.1
4
1737
XM_017026686.1
XP_016882175.1
7
2027
XM_017026687.1
XP_016882176.1
6
1889
XM_017026688.1
XP_016882177.1
4
3961
XM_017026689.1
XP_016882178.1
6
1975
XM_017026690.1
XP_016882179.1
4
1755
XM_017026691.1
XP_016882180.1
3
1535
XM_017026692.1
XP_016882181.1
8
2105
XM_017026693.1
XP_016882182.1
9
2226
XM_017026694.1
XP_016882183.1
9
2176
XM_017026695.1
XP_016882184.1
8
2148
XM_017026696.1
XP_016882185.1
7
2000
XM_017026697.1
XP_016882186.1
6
1922
XM_017026698.1
XP_016882187.1
6
1900
XM_017026699.1
XP_016882188.1
4
4736
XM_017026700.1
XP_016882189.1
6
1901
XM_017026701.1
XP_016882190.1
8
2099
XM_017026702.1
XP_016882191.1
7
2021
XM_017026703.1
XP_016882192.1
5
1798
XM_017026704.1
XP_016882193.1
5
1860
XM_017026705.1
XP_016882194.1
2
1857
XM_017026707.1
XP_016882196.1
5
2273
XM_017026708.1
XP_016882197.1
5
2276
GenBank mRNA
AB013897.1
-
5
1802
AK056243.1
-
6
1889
AK128116.1
-
9
2508
AK223616.1
-
5
1795
AK299969.1
-
5
1769
AK301601.1
-
5
1250
AK307852.1
-
1
1513
BC004513.2
-
1
977
BC017256.2
-
6
1955
3356680
BC053845.1
-
4
1792
5493685
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0