Search
Search

RPUSD3| Gene Symbol: RPUSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142547.1 | NP_001136019.1 | 6, 7 | 686 | ||
| NM_173659.3 | NP_775930.2 | 7, 8 | 731 | |||
| GenBank mRNA | AK291888.1 | - | 7, 8 | 732 | ||
| BC032135.2 | - | 7, 8 | 705 | 4649090 | ||
| BC065741.1 | - | 7, 8 | 733 | 4091801 | ||