Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164673.1 | NP_001158145.1 | 4 | 565 | ||
NM_001301071.1 | NP_001288000.1 | 4 | 565 | |||
NM_173660.4 | NP_775931.3 | 4 | 565 | |||
XM_011513435.2 | XP_011511737.1 | 4 | 565 | |||
XM_011513437.2 | XP_011511739.1 | 4 | 565 | |||
GenBank mRNA | AB220918.1 | - | 4 | 495 | ||
AK091037.1 | - | 4 | 560 | |||
BC131544.1 | - | 4 | 502 | 40085498 | ||
BC141852.1 | - | 4 | 536 | 8860352 | ||
DA205628.1 | - | 4 | 565 |