Search
Search

LURAP1L| Gene Symbol: LURAP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203403.1 | NP_981948.1 | 1 | 988 | ||
| XM_005251443.2 | XP_005251500.1 | 1 | 1011 | |||
| GenBank mRNA | AB073606.1 | - | 1 | 990 | ||
| AK074752.1 | - | 1 | 469 | |||
| AK095824.1 | - | 1 | 987 | |||
| BC036923.1 | - | 1 | 963 | 5228515 | ||
| BM739457.1 | - | 1 | 204 | |||