Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207354.2 | NP_997237.2 | 4 | 535 | ||
NR_030767.1 | 5 | 618 | ||||
XM_011544985.1 | XP_011543287.1 | 4 | 478 | |||
XM_011544986.1 | XP_011543288.1 | 5 | 618 | |||
XM_011544987.2 | XP_011543289.1 | 4 | 452 | |||
GenBank mRNA | AK096277.1 | - | 4 | 448 | ||
AK124721.1 | - | 5 | 615 | |||
AK126438.1 | - | 3 | 532 | |||
BC110419.1 | - | 4 | 508 | 5210878 | ||
BC121024.1 | - | 4 | 409 | 40117420 | ||
BC121025.2 | - | 4 | 409 | 40117423 |