Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199350.3 | NP_955382.3 | 6 | 1262 | ||
XM_011518655.1 | XP_011516957.1 | 8 | 1817 | |||
XM_011518656.1 | XP_011516958.1 | 7 | 1327 | |||
XM_011518657.2 | XP_011516959.1 | 7 | 1058 | |||
XM_011518658.2 | XP_011516960.1 | 7 | 1055 | |||
XM_011518660.1 | XP_011516962.1 | 7 | 1087 | |||
XM_011518661.2 | XP_011516963.1 | 4 | 791 | |||
XM_011518662.2 | XP_011516964.1 | 4 | 788 | |||
GenBank mRNA | AK093122.1 | - | 6 | 1262 | ||
AW468767.1 | - | 2 | 359 | |||
AW958564.1 | - | 3 | 235 | |||
AW958565.1 | - | 3 | 235 | |||
BC112353.1 | - | 4 | 482 | |||
JF442034.1 | - | 6 | 730 |