Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212551.4 | NP_997716.1 | 1 | 793 | ||
GenBank mRNA | AK298814.1 | - | 1 | 181 | ||
AY037156.1 | - | 1 | 215 | |||
BC031649.1 | - | 1 | 397 | 5172124 | ||
BX647911.1 | - | 1 | 759 |