Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302644.1 | NP_001289573.1 | 2 | 2770 | ||
NM_001302645.1 | NP_001289574.1 | 2 | 2637 | |||
NM_001302646.1 | NP_001289575.1 | 2 | 2808 | |||
NM_001302647.1 | NP_001289576.1 | 2 | 2678 | |||
NM_001302648.1 | NP_001289577.1 | 2 | 2643 | |||
NM_207429.3 | NP_997312.1 | 2 | 2852 | |||
GenBank mRNA | AB096245.1 | - | 2 | 2769 | ||
AK127703.1 | - | 1 | 495 | |||
CR936778.1 | - | 2 | 2767 | |||
KF728566.1 | - | 2 | 2770 | |||
KF728567.1 | - | 2 | 2637 | |||
KF728568.1 | - | 2 | 2852 | |||
KF728569.1 | - | 2 | 2770 | |||
KF728570.1 | - | 2 | 2808 |