Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001786.2 | NP_001001786.2 | 1 | 447 | ||
GenBank mRNA | AF303179.1 | - | 1 | 447 | ||
BC130361.1 | - | 1 | 325 | 40146392 | ||
BC130363.1 | - | 1 | 325 | 40146394 |