Search
Search

BRD4| Gene Symbol: BRD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014299.2 | NP_055114.1 | 8 | 1703 | ||
| NM_058243.2 | NP_490597.1 | 8 | 1703 | |||
| XM_011527854.1 | XP_011526156.1 | 8 | 1768 | |||
| XM_011527855.1 | XP_011526157.1 | 8 | 1769 | |||
| XM_011527856.2 | XP_011526158.1 | 8 | 1768 | |||
| GenBank mRNA | AF386649.1 | - | 8 | 1703 | ||
| AK307664.1 | - | 8 | 1593 | |||
| BC000156.1 | - | 8 | 1549 | |||
| BC030158.2 | - | 8 | 1570 | |||
| BC035266.1 | - | 8 | 1774 | 4837491 | ||
| BC038988.1 | - | 8 | 1744 | |||
| BC047888.1 | - | 8 | 1757 | |||
| BC067129.1 | - | 8 | 1725 | |||
| BC091649.1 | - | 8 | 1741 | |||
| KF974357.1 | - | 2 | 236 | |||
| Y12059.1 | - | 8 | 1703 | |||