Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004356.2 | NP_001004356.1 | 5 | 761 | ||
NM_001004358.1 | NP_001004358.1 | 5 | 644 | |||
NM_021923.3 | NP_068742.2 | 4 | 634 | |||
XM_011513486.1 | XP_011511788.1 | 5 | 765 | |||
GenBank mRNA | AF279689.1 | - | 5 | 644 | ||
AF312678.1 | - | 761 | ||||
AJ277437.1 | - | 4 | 634 | |||
AK172829.1 | - | 4 | 618 | |||
AK225486.1 | - | 4 | 605 | |||
AK314365.1 | - | 5 | 762 | |||
AY358303.1 | - | 761 | ||||
BC015228.1 | - | 1 | 45 | |||
BC036769.1 | - | 5 | 773 | 5744684 | ||
BQ689625.1 | - | 4 | 590 |