Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003952.2 | NP_003943.2 | 13 | 1146 | ||
XM_005274164.1 | XP_005274221.1 | 12 | 1094 | |||
XM_005274165.4 | XP_005274222.1 | 12 | 971 | |||
XM_006718655.3 | XP_006718718.1 | 14 | 1267 | |||
XM_006718656.3 | XP_006718719.1 | 13 | 1215 | |||
XM_006718657.1 | XP_006718720.1 | 13 | 1200 | |||
XM_017018108.1 | XP_016873597.1 | 14 | 1278 | |||
GenBank mRNA | AB016869.1 | - | 13 | 1118 | ||
AB019245.1 | - | 13 | 1113 | |||
AB208827.1 | - | 7 | 4619 | |||
AF076931.1 | - | 13 | 1099 | |||
AF099739.1 | - | 13 | 1103 | |||
BC000094.2 | - | 13 | 1119 | 3508140 | ||
BC006106.1 | - | 13 | 1102 | 2959036 | ||
BC136564.1 | - | 13 | 1120 | 9020554 | ||
BX538119.1 | - | 5 | 301 |