Search
Search

CHKA| Gene Symbol: CHKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277.2 | NP_001268.2 | 6, 7 | 1088 | ||
| NM_212469.1 | NP_997634.1 | 5, 6 | 1034 | |||
| XM_017017147.1 | XP_016872636.1 | 5, 6 | 972 | |||
| XM_017017148.1 | XP_016872637.1 | 5, 6 | 2444 | |||
| GenBank mRNA | BC036471.2 | - | 5, 6 | 912 | 5265994 | |
| D10704.1 | - | 6, 7 | 898 | |||