Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADARB1 SIRNA Assays ›
Gene Symbol:
ADARB1
Gene Name:
adenosine deaminase, RNA specific B1
Gene Aliases:
ADAR2, DRABA2, DRADA2, RED1
Chromosome Location:
Chr.21: 45074578 - 45226563 on Build GRCh38
UniGene ID:
Hs.474018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS141147
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADARB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112.3
NP_001103.1
9
2150
NM_001160230.1
NP_001153702.1
9
2150
NM_015833.3
NP_056648.1
10
2270
NM_015834.3
NP_056649.1
10
2270
NR_027672.1
8
1215
NR_027673.1
10
2268
NR_027674.1
9
2150
NR_073200.1
10
2270
XM_006723954.2
XP_006724017.1
11
2342
XM_011529425.2
XP_011527727.1
11
3462
XM_011529432.1
XP_011527734.1
9
1183
XM_017028242.1
XP_016883731.1
9
3938
XM_017028243.1
XP_016883732.1
12
7427
XM_017028244.1
XP_016883733.1
9
3936
XM_017028245.1
XP_016883734.1
8
3771
XM_017028246.1
XP_016883735.1
12
7427
XM_017028247.1
XP_016883736.1
11
7309
XM_017028248.1
XP_016883737.1
12
2251
XM_017028249.1
XP_016883738.1
13
7470
XM_017028250.1
XP_016883739.1
12
7380
XM_017028251.1
XP_016883740.1
9
2052
XM_017028252.1
XP_016883741.1
9
3938
XM_017028253.1
XP_016883742.1
8
3766
XM_017028254.1
XP_016883743.1
11
7309
XM_017028255.1
XP_016883744.1
10
7187
XM_017028256.1
XP_016883745.1
9
1982
GenBank mRNA
AB194369.1
-
8
1193
AB194370.1
-
9
2128
AB194371.1
-
9
2128
AB194372.2
-
9
2128
AB194373.1
-
9
2128
AF001042.1
-
8
1850
AF525422.1
-
10
2246
AF533142.1
-
9
2128
AY135659.1
-
10
2246
BC065545.1
-
9
1987
6014605
U76420.1
-
9
2128
U76421.1
-
10
2248
U76422.1
-
10
2248
U82120.1
-
9
1955
U82121.1
-
8
1835
X99227.1
-
9
1982
X99383.1
-
8
1862
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
adenosine to inosine editing
mRNA processing
negative regulation of cell proliferation
negative regulation of cell migration
negative regulation of protein kinase activity by regulation of protein phosphorylation
positive regulation of viral genome replication
innate immune response
defense response to virus
regulation of cell cycle
double-stranded RNA adenosine deaminase activity
mRNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0