Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADRA1A SIRNA Assays ›
          Gene Symbol:
          ADRA1A
          Gene Name:
          adrenoceptor alpha 1A
          Gene Aliases:
          ADRA1C, ADRA1L1, ALPHA1AAR
          Chromosome Location:
          Chr.8: 26738113 - 26870994 on Build GRCh38
          UniGene ID:
          Hs.52931
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS141167
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADRA1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000680.3 NP_000671.2 2 1338
          NM_001322502.1 NP_001309431.1 1 1030
          NM_001322503.1 NP_001309432.1 1 1030
          NM_001322504.1 NP_001309433.1 1 1030
          NM_033302.3 NP_150645.2 1 1030
          NM_033303.4 NP_150646.3 1 1030
          NM_033304.3 NP_150647.2 1 1030
          NR_136343.1 1 1030
          XM_006716292.3 XP_006716355.1 2 1365
          XM_006716293.3 XP_006716356.1 2 1365
          XM_011544411.2 XP_011542713.1 2 1365
          XM_011544412.2 XP_011542714.1 2 1365
          XM_017013094.1 XP_016868583.1 3 4925
          XM_017013095.1 XP_016868584.1 3 4948
          XM_017013096.1 XP_016868585.1 2 1365
          GenBank mRNA AF013261.1 - 1 544
          AF395806.1 - 1 349
          AK289548.1 - 2 1320
          AY491775.1 - 1 780
          AY491776.1 - 1 780
          AY491777.1 - 1 780
          AY491778.1 - 1 780
          AY491780.1 - 1 780
          AY491781.1 - 1 780
          BC095512.1 - 1 356 6971913
          D25235.1 - 1 780
          D32201.1 - 1 780
          D32202.1 - 1 780
          L31774.1 - 1 344
          U02569.1 - 1 768

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017191, XM_006252109, XM_006252110, XM_006252108, XM_006252107, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001271759, NM_013461, NM_001271761, NM_001271760, AK042759, and others.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of heart rate involved in baroreceptor response to increased systemic arterial blood pressure
          norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
          positive regulation of heart rate by epinephrine-norepinephrine
          positive regulation of the force of heart contraction by epinephrine-norepinephrine
          positive regulation of systemic arterial blood pressure
          apoptotic process
          smooth muscle contraction
          signal transduction
          G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          activation of phospholipase C activity
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          adult heart development
          aging
          negative regulation of cell proliferation
          response to hormone
          cell growth
          positive regulation of synaptic transmission, GABAergic
          negative regulation of Rho protein signal transduction
          organ growth
          intracellular signal transduction
          response to drug
          positive regulation of MAPK cascade
          positive regulation of action potential
          positive regulation of vasoconstriction
          positive regulation of smooth muscle contraction
          micturition
          calcium ion transport into cytosol
          positive regulation of cardiac muscle contraction
          positive regulation of ERK1 and ERK2 cascade
          adenylate cyclase-activating adrenergic receptor signaling pathway
          positive regulation of protein kinase C signaling
          pilomotor reflex
          alpha1-adrenergic receptor activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0