Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL2L1 SIRNA Assays ›
          Gene Symbol:
          BCL2L1 ABALON
          Gene Name:
          BCL2 like 1
          apoptotic BCL2L1-antisense long non-coding RNA
          Gene Aliases:
          BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52
          INXS
          Chromosome Location:
          Chr.20: 31664452 - 31723999 on Build GRCh38
          Chromosome Location:
          Chr.20: 31721507 - 31723409 on Build GRCh38
          UniGene ID:
          Hs.516966
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS141363
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCL2L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001191.3 NP_001182.1 2 839
          NM_001317919.1 NP_001304848.1 2 821
          NM_001317920.1 NP_001304849.1 2 476
          NM_001317921.1 NP_001304850.1 2 605
          NM_001322239.1 NP_001309168.1 2 494
          NM_001322240.1 NP_001309169.1 2 623
          NM_001322242.1 NP_001309171.1 2 476
          NM_138578.2 NP_612815.1 2 839
          NR_134257.1 1 340
          XM_011528964.2 XP_011527266.1 2 458
          XM_017027993.1 XP_016883482.1 2 464
          GenBank mRNA AK290968.1 - 2 609
          BC019307.1 - 2 594 2823498
          BM554265.1 - 2 578
          BT007208.1 - 1 228
          BX359456.2 - 2 511
          CR936637.1 - 2 625
          DR156871.1 - 2 385
          KU177938.1 - 1 228
          KU177939.1 - 1 228
          KU177940.1 - 1 228
          Z23115.1 - 1 362
          Z23116.1 - 1 362
          Gene Symbol: ABALON
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_131907.1 1 481
          GenBank mRNA KC505631.1 - 1 481

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          ovarian follicle development
          in utero embryonic development
          release of cytochrome c from mitochondria
          endocytosis
          mitotic cell cycle checkpoint
          germ cell development
          spermatogenesis
          cell proliferation
          positive regulation of cell proliferation
          male gonad development
          intrinsic apoptotic signaling pathway in response to DNA damage
          apoptotic mitochondrial changes
          fertilization
          suppression by virus of host apoptotic process
          response to cytokine
          growth
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          response to cycloheximide
          regulation of mitochondrial membrane permeability
          neuron apoptotic process
          regulation of mitochondrial membrane potential
          cellular process regulating host cell cycle in response to virus
          mitochondrion morphogenesis
          cellular response to amino acid stimulus
          cellular response to alkaloid
          cellular response to gamma radiation
          apoptotic process in bone marrow
          negative regulation of establishment of protein localization to plasma membrane
          negative regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway in absence of ligand
          hepatocyte apoptotic process
          negative regulation of execution phase of apoptosis
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of intrinsic apoptotic signaling pathway
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          protein kinase binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          BH3 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0