Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080513.3 | NP_001073982.3 | 2 | 271 | ||
NM_001291988.1 | NP_001278917.1 | 2 | 268 | |||
XM_005269280.4 | XP_005269337.1 | 3 | 400 | |||
GenBank mRNA | BC031569.1 | - | 2 | 219 | 5188827 | |
BC042334.1 | - | 2 | 227 | 5190234 | ||
BC137398.1 | - | 2 | 261 | 9021396 | ||
BC137403.1 | - | 2 | 261 | 9021401 | ||
BX951477.1 | - | 2 | 272 | |||
J05158.2 | - | 1 | 185 |