Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DRD4 SIRNA Assays ›
Gene Symbol:
DRD4
Gene Name:
dopamine receptor D4
Gene Aliases:
D4DR
Chromosome Location:
Chr.11: 637305 - 640706 on Build GRCh38
UniGene ID:
Hs.99922
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS141878
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DRD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000797.3
NP_000788.2
3
572
GenBank mRNA
EU432112.1
-
3
572
L12398.1
-
3
572
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012944, XM_006230521, XM_006230522, M84009.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
behavioral fear response
synaptic transmission, dopaminergic
response to amphetamine
cellular calcium ion homeostasis
negative regulation of adenylate cyclase activity
adenylate cyclase-inhibiting dopamine receptor signaling pathway
dopamine receptor signaling pathway
short-term memory
circadian rhythm
adult locomotory behavior
olfactory learning
photoperiodism
negative regulation of cAMP biosynthetic process
positive regulation of sodium:proton antiporter activity
positive regulation of kinase activity
response to histamine
social behavior
regulation of dopamine metabolic process
dopamine metabolic process
fear response
regulation of circadian rhythm
regulation of neurotransmitter secretion
behavioral response to cocaine
behavioral response to ethanol
response to steroid hormone
arachidonic acid secretion
negative regulation of protein secretion
regulation of calcium-mediated signaling
positive regulation of dopamine uptake involved in synaptic transmission
retina development in camera-type eye
inhibitory postsynaptic potential
positive regulation of penile erection
negative regulation of voltage-gated calcium channel activity
positive regulation of excitatory postsynaptic potential
dopamine neurotransmitter receptor activity, coupled via Gs
dopamine neurotransmitter receptor activity, coupled via Gi/Go
dopamine neurotransmitter receptor activity
protein binding
drug binding
potassium channel regulator activity
SH3 domain binding
dopamine binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0