Search
Search

GABRG2| Gene Symbol: GABRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000816.3 | NP_000807.2 | 9 | 1500 | ||
| NM_198903.2 | NP_944493.2 | 11 | 1644 | |||
| NM_198904.2 | NP_944494.1 | 10 | 1524 | |||
| GenBank mRNA | AK299640.1 | - | 10 | 1225 | ||
| AK315200.1 | - | 10 | 1176 | |||
| BC059389.1 | - | 10 | 1345 | 5288256 | ||
| BC069348.1 | - | 9 | 1236 | 7262321 | ||
| BC074795.2 | - | 9 | 1174 | 30915258 | ||
| CD013896.1 | - | 10 | 1302 | |||
| CD013897.1 | - | 9 | 1210 | |||
| X15376.1 | - | 9 | 1368 | |||