Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GK SIRNA Assays ›
          Gene Symbol:
          GK
          Gene Name:
          glycerol kinase
          Gene Aliases:
          GK1, GKD
          Chromosome Location:
          Chr.X: 30653348 - 30731462 on Build GRCh38
          UniGene ID:
          Hs.1466
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS142202
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000167.5 NP_000158.1 17 1582
          NM_001128127.2 NP_001121599.1 17 1582
          NM_001205019.1 NP_001191948.1 18 1600
          NM_203391.3 NP_976325.1 18 1600
          XM_005274488.4 XP_005274545.1 15 1352
          XM_006724483.2 XP_006724546.1 18 1619
          XM_006724484.2 XP_006724547.1 18 1618
          XM_006724485.2 XP_006724548.1 17 1445
          XM_006724486.3 XP_006724549.1 16 1430
          XM_011545491.2 XP_011543793.1 19 1638
          XM_011545492.1 XP_011543794.1 19 1637
          XM_011545493.2 XP_011543795.1 17 1320
          XM_011545494.2 XP_011543796.1 17 1265
          XM_017029409.1 XP_016884898.1 19 1635
          XM_017029410.1 XP_016884899.1 16 1414
          XM_017029411.1 XP_016884900.1 16 1429
          XM_017029412.1 XP_016884901.1 15 1350
          GenBank mRNA AK294304.1 - 18 1406
          AK294938.1 - 19 1635
          AK308238.1 - 15 1261
          AK313215.1 - 17 1518
          BC037549.1 - 17 1439 5213716
          BC042421.1 - 18 1601 4820945
          BC071595.1 - 18 1601 5276056
          BX647604.1 - 18 1421
          CD369459.1 - 4 459
          L13943.1 - 17 1469
          X68285.1 - 16 1192
          X69886.1 - 13 1018

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerol metabolic process
          triglyceride metabolic process
          response to cold
          phosphorylation
          regulation of fatty acid metabolic process
          triglyceride biosynthetic process
          glycerol catabolic process
          response to drug
          glucose homeostasis
          response to ethanol
          glycerol-3-phosphate biosynthetic process
          glycerol kinase activity
          protein binding
          ATP binding
          histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-nkbdw:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline