Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPR25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005298.3 | NP_005289.2 | 1 | 701 | ||
GenBank mRNA | AK315339.1 | - | 1 | 675 | ||
BC126386.1 | - | 1 | 701 | 8992102 | ||
BC126388.1 | - | 1 | 701 | 8992104 |
Set Membership: |
G-protein coupled receptor |