Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-B SIRNA Assays ›
          Gene Symbol:
          HLA-B HLA-C
          Gene Name:
          major histocompatibility complex, class I, B
          major histocompatibility complex, class I, C
          Gene Aliases:
          AS, B-4901, B-5001, Bw-47, Bw-50, HEL-S-83, HLA-B*45ZJ, HLA-B-3506, HLA-B-3905, HLA-B-5502, HLA-B-5602, HLA-B15, HLA-B39, HLA-B49, HLA-B50, HLA-B55, HLA-B59, HLA-B61, HLA-Cw, HLA-DRB1, HLAB, SPDA1
          D6S204, HLA-JY3, HLAC, HLC-C, MHC, PSORS1
          Chromosome Location:
          Chr.6: 31353866 - 31357245 on Build GRCh38
          Chromosome Location:
          Chr.6: 31268749 - 31272136 on Build GRCh38
          UniGene ID:
          Hs.654404
          Hs.656020
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS142418
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005514.7 NP_005505.2 4 835
          XM_011514557.1 XP_011512859.1 4 758
          GenBank mRNA AB008102.1 - 4 748
          AF004370.1 - 4 652
          AF022783.1 - 4 748
          AF026218.1 - 4 748
          AF170577.1 - 4 748
          AF189017.1 - 4 748
          AJ506054.1 - 1 129
          AJ634528.1 - 4 748
          AK124160.1 - 4 1018
          AK130137.1 - 4 769
          AK130166.1 - 4 769
          AK130169.1 - 4 775
          AK130654.1 - 5 708
          AK292226.1 - 4 769
          AK300685.1 - 5 678
          AK313911.1 - 4 769
          AL576516.3 - 5 740
          AY007140.1 - 4 768
          AY102648.1 - 3 673
          AY569160.1 - 3 675
          BC007243.1 - 5 742
          BC091497.1 - 4 752 30386550
          BX647315.1 - 5 770
          D14343.1 - 4 712
          D31816.1 - 4 748
          D50290.1 - 4 748
          D50291.1 - 4 748
          D50299.1 - 4 748
          D50300.1 - 4 748
          D83043.1 - 4 748
          D85761.1 - 5 748
          DQ336696.1 - 4 736
          DQ336697.1 - 4 742
          EU048339.1 - 1 129
          EU668336.1 - 4 748
          FM205711.1 - 4 748
          HE663398.1 - 5 697
          HG327115.1 - 4 748
          HG327117.1 - 4 748
          HG327119.1 - 4 748
          HG327120.1 - 4 748
          HG514457.1 - 4 748
          KU204955.1 - 2 405
          L04695.1 - 4 748
          L04696.1 - 4 748
          L07743.1 - 4 748
          L09736.1 - 4 748
          L19923.1 - 4 748
          L20089.1 - 4 743
          L20090.1 - 5 743
          L33922.1 - 4 748
          L37880.1 - 4 748
          L38504.1 - 4 748
          L41214.1 - 5 748
          L41925.1 - 5 748
          M12678.1 - 4 739
          M16102.1 - 4 755
          M27540.1 - 3 562
          M28204.1 - 4 712
          M28205.1 - 5 748
          M32317.1 - 4 748
          M32319.1 - 5 748
          M62852.1 - 3 676
          M77774.1 - 5 748
          M77775.1 - 4 748
          M77776.1 - 4 748
          M77777.1 - 4 748
          M77778.1 - 4 748
          M81798.1 - 4 748
          M84380.1 - 4 748
          M84381.1 - 4 748
          M84383.1 - 4 748
          M84384.1 - 4 748
          M84385.1 - 4 748
          M84694.1 - 4 748
          M86403.1 - 4 724
          U03698.1 - 4 748
          U04244.1 - 4 748
          U04245.1 - 4 748
          U06697.1 - 3 675
          U06862.1 - 4 748
          U09912.1 - 4 748
          U11265.1 - 4 748
          U14943.1 - 4 748
          U17107.1 - 4 748
          U17572.1 - 4 748
          U21052.1 - 4 748
          U21053.1 - 4 748
          U29057.1 - 4 748
          U30904.1 - 4 748
          U30936.1 - 4 748
          U31971.1 - 4 748
          U35734.1 - 4 748
          U63653.1 - 4 748
          U70529.1 - 4 748
          U74386.1 - 3 652
          U88407.1 - 4 748
          X61708.1 - 5 748
          X64454.1 - 4 748
          X73578.1 - 4 748
          X81363.1 - 4 748
          X84725.1 - 4 748
          X90390.1 - 4 748
          X91749.1 - 4 748
          Y08994.1 - 5 748
          Y09118.1 - 4 748
          Y13567.1 - 4 748
          Z15143.1 - 5 748
          Z22651.1 - 4 748
          Z33453.1 - 4 749
          Gene Symbol: HLA-C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC008457.1 - 5 758 4249847
          D50853.1 - 5 748
          D64150.1 - 5 748
          HG974570.1 - 5 748
          M84172.1 - 5 748
          M99389.1 - 5 748
          M99390.1 - 5 748

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
          viral process
          antigen processing and presentation
          regulation of immune response
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          antigen binding
          peptide antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0