Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-H SIRNA Assays ›
          Gene Symbol:
          HLA-H HLA-A HLA-B
          Gene Name:
          major histocompatibility complex, class I, H (pseudogene)
          major histocompatibility complex, class I, A
          major histocompatibility complex, class I, B
          Gene Aliases:
          HLAHP
          Aw-33, Aw-74, HLA-A11, HLA-A33, HLA-DQB1, HLA-DRB1, HLAA
          AS, B-4901, B-5001, Bw-47, Bw-50, HEL-S-83, HLA-B*45ZJ, HLA-B-3506, HLA-B-3905, HLA-B-5502, HLA-B-5602, HLA-B15, HLA-B39, HLA-B49, HLA-B50, HLA-B55, HLA-B59, HLA-B61, HLA-Cw, HLA-DRB1, HLAB, SPDA1
          Chromosome Location:
          Chr.6: 29887760 - 29891080 on Build GRCh38
          Chromosome Location:
          Chr.6: 29942470 - 29945884 on Build GRCh38
          Chromosome Location:
          Chr.6: 31353866 - 31357245 on Build GRCh38
          UniGene ID:
          Hs.728808
          Hs.181244
          Hs.654404
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID HSS142450
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_001434.3 2 243
          GenBank mRNA AK090500.1 - 2 285
          AK299998.1 - 2 424
          Gene Symbol: HLA-A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AJ001274.1 - 1 178
          AK125608.1 - 2 273
          AY225407.1 - 2 251
          BC008611.1 - 2 256 4157200
          D14350.1 - 2 251
          D14351.1 - 2 251
          D14354.1 - 2 251
          D16843.1 - 2 251
          D32129.1 - 2 256
          D32130.1 - 2 304
          D32131.1 - 2 304
          D50068.1 - 2 251
          DQ327719.1 - 2 251
          DQ327720.1 - 2 251
          M24095.1 - 1 173
          M32321.1 - 2 251
          M98453.1 - 2 251
          U03697.1 - 2 251
          U41057.1 - 2 251
          U43334.1 - 1 178
          U43335.1 - 1 178
          U43336.1 - 1 178
          U45480.1 - 2 251
          U52429.1 - 2 251
          X61704.1 - 2 251
          X61711.1 - 2 251
          X96638.1 - 1 178
          X97802.1 - 2 251
          Gene Symbol: HLA-B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005514.7 NP_005505.2 2 338
          XM_011514557.1 XP_011512859.1 2 261
          GenBank mRNA AF004370.1 - 1 155
          AF016641.1 - 2 251
          AF033351.1 - 1 178
          AF170577.1 - 2 251
          AF189017.1 - 2 251
          AF298582.1 - 1 178
          AF436091.1 - 2 240
          AF436092.1 - 2 240
          AF436093.1 - 2 240
          AF436094.1 - 2 240
          AF436098.1 - 2 251
          AJ002675.1 - 1 178
          AJ002676.1 - 1 178
          AJ002677.1 - 1 178
          AJ003063.1 - 1 178
          AJ223602.1 - 1 178
          AJ276427.1 - 1 179
          AK130137.1 - 2 272
          AK130155.1 - 2 274
          AK130654.1 - 2 250
          AK292226.1 - 2 272
          AK300685.1 - 3 181
          AK301008.1 - 2 158
          AK303541.1 - 1 400
          AK310586.1 - 3 191
          AK313911.1 - 2 272
          AY007140.1 - 2 271
          AY040668.1 - 1 178
          AY102648.1 - 1 176
          AY569160.1 - 1 178
          BC007243.1 - 2 245
          BC013187.1 - 2 260 4344323
          BC091497.1 - 2 255 30386550
          BX647315.1 - 2 273
          D25275.1 - 2 251
          D50293.1 - 2 251
          D50294.1 - 2 251
          D50295.1 - 2 251
          D50296.1 - 2 251
          D50299.1 - 2 251
          D50300.1 - 2 251
          D85761.1 - 2 251
          D85762.1 - 2 251
          DA561140.1 - 3 531
          DC345094.1 - 2 305
          DQ336698.1 - 2 239
          FM205711.1 - 2 251
          HE663398.1 - 2 200
          HG327117.1 - 2 251
          HG327119.1 - 2 251
          HG327120.1 - 2 251
          HG514457.1 - 2 251
          HM989017.1 - 1 178
          L04695.1 - 2 251
          L04696.1 - 2 251
          L07743.1 - 2 251
          L07950.1 - 2 251
          L11570.1 - 2 251
          L11571.1 - 2 251
          L11604.1 - 2 251
          L11666.1 - 2 251
          L15005.1 - 2 251
          L22027.1 - 2 251
          L24373.1 - 2 251
          L32862.1 - 2 251
          L33922.1 - 2 251
          L37880.1 - 2 251
          L37881.1 - 2 251
          L41214.1 - 2 251
          L41925.1 - 2 251
          L42345.1 - 2 251
          L42545.1 - 1 178
          L47338.1 - 1 178
          L47985.1 - 1 178
          L47986.1 - 1 178
          L49341.1 - 1 178
          L49342.1 - 1 178
          L49343.1 - 1 178
          L63544.1 - 1 178
          L76930.1 - 1 178
          L76932.1 - 1 178
          L76933.1 - 1 178
          M16102.1 - 2 258
          M27540.1 - 1 65
          M28204.1 - 2 215
          M28205.1 - 2 251
          M32317.1 - 2 251
          M32319.1 - 2 251
          M75138.1 - 2 251
          M77774.1 - 3 251
          M77775.1 - 2 251
          M77776.1 - 2 251
          M77777.1 - 2 251
          M77778.1 - 2 251
          M81798.1 - 2 251
          M83192.1 - 1 179
          M84381.1 - 2 251
          M84385.1 - 2 251
          M86403.1 - 2 227
          U04244.1 - 2 251
          U04245.1 - 2 251
          U04787.1 - 2 251
          U06697.1 - 1 178
          U09864.1 - 2 251
          U09912.1 - 2 251
          U11262.1 - 2 251
          U11264.1 - 2 251
          U11265.1 - 2 251
          U11266.1 - 2 251
          U11268.1 - 2 251
          U11269.1 - 2 251
          U17107.1 - 2 251
          U18661.1 - 1 35
          U18789.1 - 1 178
          U21052.1 - 2 251
          U21053.1 - 2 251
          U28759.1 - 1 158
          U29057.1 - 2 251
          U30904.1 - 2 251
          U30936.1 - 2 251
          U31334.1 - 1 178
          U32661.1 - 1 178
          U32678.1 - 1 178
          U34618.1 - 1 178
          U34619.1 - 1 178
          U34810.1 - 1 178
          U36392.1 - 2 251
          U36492.1 - 2 251
          U38800.1 - 2 251
          U57966.1 - 2 251
          U74386.1 - 1 155
          U80945.1 - 2 251
          U88407.1 - 2 251
          X61708.1 - 2 251
          X64454.1 - 2 251
          X77658.1 - 2 251
          X90390.1 - 2 251
          X91749.1 - 2 251
          X96473.1 - 1 149
          Y07710.1 - 1 178
          Y07824.1 - 1 178
          Y08994.1 - 2 251
          Y09058.1 - 2 251
          Y13567.1 - 2 251
          Z15143.1 - 2 251
          Z22651.1 - 2 251
          Z46808.1 - 2 251

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
          immune response
          antigen processing and presentation
          regulation of immune response
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          viral process
          antigen binding
          beta-2-microglobulin binding
          peptide antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0